Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610800 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATG10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global - Not Available | Caucasian
|
CEPH (CEU)
|
||||||
EAS - Not Available | African American
|
YRI (Yoruba)
|
||||||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
||||||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR - Not Available | ||||||||
AMR - Not Available |
ATG10 - autophagy related 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001131028.1 | 557 | Intron | NP_001124500.1 | |||
NM_031482.4 | 557 | Intron | NP_113670.1 | |||
XM_005248610.4 | 557 | UTR 5 | XP_005248667.1 | |||
XM_005248611.4 | 557 | Intron | XP_005248668.1 | |||
XM_005248612.3 | 557 | Intron | XP_005248669.1 | |||
XM_011543660.2 | 557 | Intron | XP_011541962.1 | |||
XM_011543661.2 | 557 | Intron | XP_011541963.1 | |||
XM_017009944.1 | 557 | Intron | XP_016865433.1 |
Set Membership: |
HapMap Validated |