Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 138440 | ||||||||||||||||||||
Literature Links: |
GART PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GART - phosphoribosylglycinamide formyltransferase, phosphoribosylglycinamide synthetase, phosphoribosylaminoimidazole synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000819.4 | 3188 | Missense Mutation | GAA,GAT | E,D 959 | NP_000810.1 | |
NM_001136005.1 | 3188 | Missense Mutation | GAA,GAT | E,D 959 | NP_001129477.1 | |
NM_001136006.1 | 3188 | Missense Mutation | GAA,GAT | E,D 959 | NP_001129478.1 | |
NM_175085.2 | 3188 | Intron | NP_780294.1 | |||
XM_005260941.1 | 3188 | Missense Mutation | GAA,GAT | E,D 959 | XP_005260998.1 | |
XM_006723989.1 | 3188 | Missense Mutation | GAA,GAT | E,D 959 | XP_006724052.1 | |
XM_006723990.1 | 3188 | Missense Mutation | GAA,GAT | E,D 959 | XP_006724053.1 | |
XM_011529526.1 | 3188 | Missense Mutation | GAA,GAT | E,D 959 | XP_011527828.1 |