Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608369 | ||||||||||||||||||||
Literature Links: |
GALNT13 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GALNT13 - polypeptide N-acetylgalactosaminyltransferase 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301627.1 | Intron | NP_001288556.1 | ||||
NM_052917.3 | Intron | NP_443149.2 | ||||
XM_011510537.2 | Intron | XP_011508839.1 | ||||
XM_011510538.2 | Intron | XP_011508840.1 | ||||
XM_017003258.1 | Intron | XP_016858747.1 | ||||
XM_017003259.1 | Intron | XP_016858748.1 | ||||
XM_017003260.1 | Intron | XP_016858749.1 | ||||
XM_017003261.1 | Intron | XP_016858750.1 | ||||
XM_017003262.1 | Intron | XP_016858751.1 | ||||
XM_017003263.1 | Intron | XP_016858752.1 |