Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607059 MIM: 604546 MIM: 611952 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC39A4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC39A4 - solute carrier family 39 member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TONSL - tonsoku-like, DNA repair protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS28 - VPS28, ESCRT-I subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016208.3 | Intron | NP_057292.1 | ||||
NM_183057.2 | Intron | NP_898880.1 | ||||
XM_005272323.3 | Intron | XP_005272380.1 | ||||
XM_005272324.3 | Intron | XP_005272381.1 | ||||
XM_005272325.3 | Intron | XP_005272382.1 | ||||
XM_011517099.1 | Intron | XP_011515401.1 | ||||
XM_011517100.1 | Intron | XP_011515402.1 | ||||
XM_017013529.1 | Intron | XP_016869018.1 |
Set Membership: |
HapMap |