Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF419 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF419 - zinc finger protein 419 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098491.1 | Intron | NP_001091961.1 | ||||
NM_001098492.1 | Intron | NP_001091962.1 | ||||
NM_001098493.1 | Intron | NP_001091963.1 | ||||
NM_001098494.1 | Intron | NP_001091964.1 | ||||
NM_001098495.1 | Intron | NP_001091965.1 | ||||
NM_001098496.1 | Intron | NP_001091966.1 | ||||
NM_001291743.1 | Intron | NP_001278672.1 | ||||
NM_001291744.1 | Intron | NP_001278673.1 | ||||
NM_001291745.1 | Intron | NP_001278674.1 | ||||
NM_024691.3 | Intron | NP_078967.3 |
ZNF773 - zinc finger protein 773 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |