Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606618 MIM: 607291 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUSP14 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DUSP14 - dual specificity phosphatase 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYNRG - synergin, gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163544.2 | 4077 | Silent Mutation | CAC,CAT | H,H 1212 | NP_001157016.1 | |
NM_001163545.2 | 4077 | Silent Mutation | CAC,CAT | H,H 1211 | NP_001157017.1 | |
NM_001163546.2 | 4077 | Silent Mutation | CAC,CAT | H,H 1155 | NP_001157018.1 | |
NM_001163547.2 | 4077 | Silent Mutation | CAC,CAT | H,H 1084 | NP_001157019.1 | |
NM_007247.5 | 4077 | Silent Mutation | CAC,CAT | H,H 1290 | NP_009178.3 | |
NM_080550.4 | 4077 | Silent Mutation | CAC,CAT | H,H 1200 | NP_542117.3 | |
NM_198882.2 | 4077 | Silent Mutation | CAC,CAT | H,H 1235 | NP_942583.1 | |
XM_005256980.4 | 4077 | Silent Mutation | CAC,CAT | H,H 1391 | XP_005257037.1 | |
XM_017024076.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1414 | XP_016879565.1 | |
XM_017024077.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1413 | XP_016879566.1 | |
XM_017024078.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1402 | XP_016879567.1 | |
XM_017024079.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1393 | XP_016879568.1 | |
XM_017024080.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1387 | XP_016879569.1 | |
XM_017024081.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1386 | XP_016879570.1 | |
XM_017024082.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1379 | XP_016879571.1 | |
XM_017024083.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1369 | XP_016879572.1 | |
XM_017024084.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1368 | XP_016879573.1 | |
XM_017024085.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1358 | XP_016879574.1 | |
XM_017024086.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1346 | XP_016879575.1 | |
XM_017024087.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1345 | XP_016879576.1 | |
XM_017024088.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1336 | XP_016879577.1 | |
XM_017024089.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1334 | XP_016879578.1 | |
XM_017024090.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1313 | XP_016879579.1 | |
XM_017024091.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1312 | XP_016879580.1 | |
XM_017024092.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1312 | XP_016879581.1 | |
XM_017024093.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1278 | XP_016879582.1 | |
XM_017024094.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1277 | XP_016879583.1 | |
XM_017024095.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1269 | XP_016879584.1 | |
XM_017024096.1 | 4077 | Intron | XP_016879585.1 | |||
XM_017024097.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1245 | XP_016879586.1 | |
XM_017024098.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1244 | XP_016879587.1 | |
XM_017024099.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1234 | XP_016879588.1 | |
XM_017024100.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1232 | XP_016879589.1 | |
XM_017024101.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1231 | XP_016879590.1 | |
XM_017024102.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1199 | XP_016879591.1 | |
XM_017024103.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1189 | XP_016879592.1 | |
XM_017024104.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1166 | XP_016879593.1 | |
XM_017024105.1 | 4077 | Intron | XP_016879594.1 | |||
XM_017024106.1 | 4077 | Silent Mutation | CAC,CAT | H,H 1167 | XP_016879595.1 |