Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615178 MIM: 191321 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KXD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KXD1 - KxDL motif containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171948.1 | Intron | NP_001165419.1 | ||||
NM_001171949.1 | Intron | NP_001165420.1 | ||||
NM_024069.3 | Intron | NP_076974.2 | ||||
XM_005260073.4 | Intron | XP_005260130.1 | ||||
XM_006722883.2 | Intron | XP_006722946.1 | ||||
XM_011528272.2 | Intron | XP_011526574.1 | ||||
XM_011528273.2 | Intron | XP_011526575.1 | ||||
XM_017027281.1 | Intron | XP_016882770.1 | ||||
XM_017027282.1 | Intron | XP_016882771.1 |
UBA52 - ubiquitin A-52 residue ribosomal protein fusion product 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |