Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608159 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRSS21 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PRSS21 - protease, serine 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270452.1 | Intron | NP_001257381.1 | ||||
NM_006799.3 | Intron | NP_006790.1 | ||||
NM_144956.2 | Intron | NP_659205.1 | ||||
NM_144957.2 | Intron | NP_659206.1 | ||||
XM_017022875.1 | Intron | XP_016878364.1 | ||||
XM_017022876.1 | Intron | XP_016878365.1 | ||||
XM_017022877.1 | Intron | XP_016878366.1 | ||||
XM_017022878.1 | Intron | XP_016878367.1 |
Set Membership: |
HapMap |