Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611128 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MDGA2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MDGA2 - MAM domain containing glycosylphosphatidylinositol anchor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113498.2 | 4967 | Intron | NP_001106970.3 | |||
NM_182830.4 | 4967 | Intron | NP_878250.2 | |||
XM_011536519.2 | 4967 | UTR 3 | XP_011534821.1 | |||
XM_011536522.2 | 4967 | Intron | XP_011534824.1 | |||
XM_011536525.2 | 4967 | UTR 3 | XP_011534827.1 | |||
XM_017021060.1 | 4967 | UTR 3 | XP_016876549.1 | |||
XM_017021061.1 | 4967 | Intron | XP_016876550.1 |
Set Membership: |
HapMap |