Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610522 MIM: 606198 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5orf38 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
C5orf38 - chromosome 5 open reading frame 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001294337.1 | Intron | NP_001281266.1 | ||||
NM_001306149.1 | Intron | NP_001293078.1 | ||||
NM_001306150.1 | Intron | NP_001293079.1 | ||||
NM_178569.3 | Intron | NP_848664.1 | ||||
XM_005248256.3 | Intron | XP_005248313.1 | ||||
XM_005248257.3 | Intron | XP_005248314.1 | ||||
XM_011513975.2 | Intron | XP_011512277.1 | ||||
XM_011513977.2 | Intron | XP_011512279.1 |
IRX2 - iroquois homeobox 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |