Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186720 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CD6 - CD6 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001254750.1 | Intron | NP_001241679.1 | ||||
NM_001254751.1 | Intron | NP_001241680.1 | ||||
NM_006725.4 | Intron | NP_006716.3 | ||||
XM_006718738.1 | Intron | XP_006718801.1 | ||||
XM_006718739.1 | Intron | XP_006718802.1 | ||||
XM_006718740.1 | Intron | XP_006718803.1 | ||||
XM_006718741.1 | Intron | XP_006718804.1 | ||||
XM_011545360.1 | Intron | XP_011543662.1 | ||||
XM_011545362.1 | Intron | XP_011543664.1 |