Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610894 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NRG4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NRG4 - neuregulin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138573.3 | 776 | Intron | NP_612640.1 | |||
XM_017021937.1 | 776 | Intron | XP_016877426.1 | |||
XM_017021938.1 | 776 | Intron | XP_016877427.1 | |||
XM_017021939.1 | 776 | Intron | XP_016877428.1 | |||
XM_017021940.1 | 776 | Intron | XP_016877429.1 | |||
XM_017021941.1 | 776 | Intron | XP_016877430.1 | |||
XM_017021942.1 | 776 | Intron | XP_016877431.1 | |||
XM_017021943.1 | 776 | Intron | XP_016877432.1 | |||
XM_017021944.1 | 776 | Intron | XP_016877433.1 | |||
XM_017021945.1 | 776 | Intron | XP_016877434.1 | |||
XM_017021946.1 | 776 | Intron | XP_016877435.1 | |||
XM_017021947.1 | 776 | Intron | XP_016877436.1 | |||
XM_017021948.1 | 776 | UTR 5 | XP_016877437.1 |
TMEM266 - transmembrane protein 266 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152335.2 | 776 | Intron | NP_689548.2 | |||
XM_005254160.2 | 776 | Intron | XP_005254217.1 | |||
XM_017021915.1 | 776 | Intron | XP_016877404.1 | |||
XM_017021916.1 | 776 | Intron | XP_016877405.1 | |||
XM_017021917.1 | 776 | Intron | XP_016877406.1 |