Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600242 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCR7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCR7 - C-C motif chemokine receptor 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301714.1 | 990 | Silent Mutation | CAA,CAG | Q,Q 240 | NP_001288643.1 | |
NM_001301716.1 | 990 | Silent Mutation | CAA,CAG | Q,Q 297 | NP_001288645.1 | |
NM_001301717.1 | 990 | Silent Mutation | CAA,CAG | Q,Q 297 | NP_001288646.1 | |
NM_001301718.1 | 990 | Silent Mutation | CAA,CAG | Q,Q 297 | NP_001288647.1 | |
NM_001838.3 | 990 | Silent Mutation | CAA,CAG | Q,Q 303 | NP_001829.1 |