Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 138033 | ||||||||||||||||||||
Literature Links: |
GCGR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GCGR - glucagon receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000160.4 | 545 | Missense Mutation | GAC,TAC | D,Y 84 | NP_000151.1 | |
XM_006722277.1 | 545 | Missense Mutation | GAC,TAC | D,Y 84 | XP_006722340.1 | |
XM_011523539.1 | 545 | Intron | XP_011521841.1 | |||
XM_017024446.1 | 545 | Missense Mutation | GAC,TAC | D,Y 82 | XP_016879935.1 | |
XM_017024447.1 | 545 | UTR 5 | XP_016879936.1 |