Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615526 MIM: 163260 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COPZ2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COPZ2 - coatomer protein complex subunit zeta 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NFE2L1 - nuclear factor, erythroid 2 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003204.2 | 838 | Missense Mutation | CAT,GAT | H,D 63 | NP_003195.1 | |
XM_005257410.3 | 838 | Missense Mutation | CAT,GAT | H,D 63 | XP_005257467.1 | |
XM_005257411.3 | 838 | Missense Mutation | CAT,GAT | H,D 63 | XP_005257468.1 | |
XM_005257412.3 | 838 | Missense Mutation | CAT,GAT | H,D 63 | XP_005257469.1 | |
XM_005257413.3 | 838 | Missense Mutation | CAT,GAT | H,D 63 | XP_005257470.1 | |
XM_005257415.3 | 838 | Missense Mutation | CAT,GAT | H,D 63 | XP_005257472.1 | |
XM_017024691.1 | 838 | Missense Mutation | CAT,GAT | H,D 63 | XP_016880180.1 |