Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 165161 MIM: 600538 MIM: 606034 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
JUNB PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
JUNB - JunB proto-oncogene, AP-1 transcription factor subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002229.2 | 750 | Silent Mutation | TCA,TCC | S,S 158 | NP_002220.1 |
MIR5684 - microRNA 5684 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRDX2 - peroxiredoxin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNASEH2A - ribonuclease H2 subunit A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |