Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179530 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAP1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAP1B - RAP1B, member of RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010942.2 | Intron | NP_001010942.1 | ||||
NM_001251917.1 | Intron | NP_001238846.1 | ||||
NM_001251918.1 | Intron | NP_001238847.1 | ||||
NM_001251921.1 | Intron | NP_001238850.1 | ||||
NM_001251922.1 | Intron | NP_001238851.1 | ||||
NM_015646.5 | Intron | NP_056461.1 |