Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606379 MIM: 611318 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR87 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GPR87 - G protein-coupled receptor 87 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023915.3 | 898 | Silent Mutation | CTA,CTG | L,L 179 | NP_076404.3 |
MED12L - mediator complex subunit 12 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_053002.5 | 898 | Intron | NP_443728.3 | |||
XM_006713487.3 | 898 | Intron | XP_006713550.1 | |||
XM_011512390.2 | 898 | Intron | XP_011510692.1 | |||
XM_011512394.2 | 898 | Intron | XP_011510696.1 | |||
XM_011512399.2 | 898 | Intron | XP_011510701.1 | |||
XM_017005676.1 | 898 | Intron | XP_016861165.1 | |||
XM_017005677.1 | 898 | Intron | XP_016861166.1 | |||
XM_017005678.1 | 898 | Intron | XP_016861167.1 | |||
XM_017005679.1 | 898 | Intron | XP_016861168.1 | |||
XM_017005680.1 | 898 | Intron | XP_016861169.1 | |||
XM_017005681.1 | 898 | Intron | XP_016861170.1 |
Set Membership: |
HapMap Validated |