Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612887 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEPT11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEPT11 - septin 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306147.1 | Intron | NP_001293076.1 | ||||
NM_018243.3 | Intron | NP_060713.1 | ||||
XM_006714259.3 | Intron | XP_006714322.1 | ||||
XM_017008400.1 | Intron | XP_016863889.1 | ||||
XM_017008401.1 | Intron | XP_016863890.1 | ||||
XM_017008402.1 | Intron | XP_016863891.1 | ||||
XM_017008403.1 | Intron | XP_016863892.1 | ||||
XM_017008404.1 | Intron | XP_016863893.1 |