Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613401 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOXRED1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NOXRED1 - NADP dependent oxidoreductase domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VIPAS39 - VPS33B interacting protein, apical-basolateral polarity regulator, spe-39 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193314.1 | Intron | NP_001180243.1 | ||||
NM_001193315.1 | Intron | NP_001180244.1 | ||||
NM_001193316.1 | Intron | NP_001180245.1 | ||||
NM_001193317.1 | Intron | NP_001180246.1 | ||||
NM_022067.3 | Intron | NP_071350.2 | ||||
XM_011537066.1 | Intron | XP_011535368.1 | ||||
XM_017021579.1 | Intron | XP_016877068.1 | ||||
XM_017021580.1 | Intron | XP_016877069.1 | ||||
XM_017021581.1 | Intron | XP_016877070.1 |