Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607593 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MDC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MDC1 - mediator of DNA damage checkpoint 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014641.2 | Intron | NP_055456.2 | ||||
XM_005249492.2 | Intron | XP_005249549.1 | ||||
XM_005249493.3 | Intron | XP_005249550.1 | ||||
XM_005249494.4 | Intron | XP_005249551.1 | ||||
XM_005249497.3 | Intron | XP_005249554.1 | ||||
XM_005249498.3 | Intron | XP_005249555.1 | ||||
XM_011515001.2 | Intron | XP_011513303.1 | ||||
XM_011515003.2 | Intron | XP_011513305.1 | ||||
XM_011515004.2 | Intron | XP_011513306.1 | ||||
XM_017011519.1 | Intron | XP_016867008.1 | ||||
XM_017011520.1 | Intron | XP_016867009.1 | ||||
XM_017011521.1 | Intron | XP_016867010.1 | ||||
XM_017011522.1 | Intron | XP_016867011.1 |
MDC1-AS1 - MDC1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |