Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612969 MIM: 601473 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TIGD7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TIGD7 - tigger transposable element derived 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF75A - zinc finger protein 75a | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302109.1 | 370 | Silent Mutation | TAC,TAT | Y,Y 180 | NP_001289038.1 | |
NM_001324041.1 | 370 | Silent Mutation | TAC,TAT | Y,Y 180 | NP_001310970.1 | |
NM_001324085.1 | 370 | Silent Mutation | TAC,TAT | Y,Y 180 | NP_001311014.1 | |
NM_153028.3 | 370 | UTR 5 | NP_694573.1 | |||
XM_011522646.2 | 370 | Silent Mutation | TAC,TAT | Y,Y 180 | XP_011520948.1 | |
XM_011522647.2 | 370 | UTR 5 | XP_011520949.1 | |||
XM_017023642.1 | 370 | UTR 5 | XP_016879131.1 | |||
XM_017023643.1 | 370 | Silent Mutation | TAC,TAT | Y,Y 180 | XP_016879132.1 | |
XM_017023644.1 | 370 | Silent Mutation | TAC,TAT | Y,Y 180 | XP_016879133.1 | |
XM_017023645.1 | 370 | Silent Mutation | TAC,TAT | Y,Y 180 | XP_016879134.1 |
Set Membership: |
HapMap Validated |