Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGAGCGGTGCCCGGGCCCGCTTAC[C/G]TGCACCGCTTGCTCCGAGCCGCGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604160 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITGB8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ITGB8 - integrin subunit beta 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002214.2 | 549 | UTR 5 | NP_002205.1 | |||
XM_011515393.2 | 549 | Intron | XP_011513695.1 | |||
XM_011515394.2 | 549 | Intron | XP_011513696.1 | |||
XM_011515396.1 | 549 | Intron | XP_011513698.1 | |||
XM_017012178.1 | 549 | UTR 5 | XP_016867667.1 | |||
XM_017012179.1 | 549 | UTR 5 | XP_016867668.1 | |||
XM_017012180.1 | 549 | UTR 5 | XP_016867669.1 | |||
XM_017012181.1 | 549 | Intron | XP_016867670.1 | |||
XM_017012182.1 | 549 | Intron | XP_016867671.1 | |||
XM_017012183.1 | 549 | Intron | XP_016867672.1 |
LOC101927811 - uncharacterized LOC101927811 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |