Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602034 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRHR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CRHR2 - corticotropin releasing hormone receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202475.1 | 2236 | Silent Mutation | NP_001189404.1 | |||
NM_001202481.1 | 2236 | Silent Mutation | NP_001189410.1 | |||
NM_001202482.1 | 2236 | Silent Mutation | NP_001189411.1 | |||
NM_001202483.1 | 2236 | Silent Mutation | NP_001189412.1 | |||
NM_001883.4 | 2236 | Silent Mutation | NP_001874.2 | |||
XM_011515127.2 | 2236 | Missense Mutation | XP_011513429.1 | |||
XM_011515128.2 | 2236 | Silent Mutation | XP_011513430.1 | |||
XM_011515129.2 | 2236 | Silent Mutation | XP_011513431.1 | |||
XM_017011751.1 | 2236 | Missense Mutation | XP_016867240.1 | |||
XM_017011752.1 | 2236 | Silent Mutation | XP_016867241.1 |
Set Membership: |
HapMap |