Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605101 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TAB2 - TGF-beta activated kinase 1/MAP3K7 binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001292034.2 | 176 | Intron | NP_001278963.1 | |||
NM_001292035.2 | 176 | UTR 5 | NP_001278964.1 | |||
NM_015093.5 | 176 | Intron | NP_055908.1 | |||
XM_011535633.2 | 176 | Intron | XP_011533935.1 | |||
XM_017010591.1 | 176 | Intron | XP_016866080.1 | |||
XM_017010592.1 | 176 | Intron | XP_016866081.1 |