Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607477 MIM: 610230 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTSE1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GTSE1 - G2 and S-phase expressed 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRMU - tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282782.1 | 439 | UTR 5 | NP_001269711.1 | |||
NM_001282783.1 | 439 | UTR 5 | NP_001269712.1 | |||
NM_001282784.1 | 439 | UTR 5 | NP_001269713.1 | |||
NM_001282785.1 | 439 | Silent Mutation | AGG,AGT | R,S 25 | NP_001269714.1 | |
NM_018006.4 | 439 | Silent Mutation | AGG,AGT | R,S 25 | NP_060476.2 | |
XM_005261678.1 | 439 | UTR 5 | XP_005261735.1 | |||
XM_005261681.1 | 439 | UTR 5 | XP_005261738.1 | |||
XM_011530271.2 | 439 | Intron | XP_011528573.1 | |||
XM_011530272.1 | 439 | Silent Mutation | AGG,AGT | R,S 25 | XP_011528574.1 | |
XM_011530273.1 | 439 | Silent Mutation | AGG,AGT | R,S 25 | XP_011528575.1 | |
XM_011530274.2 | 439 | Intron | XP_011528576.1 | |||
XM_011530275.1 | 439 | Intron | XP_011528577.1 |
Set Membership: |
HapMap |