Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 109560 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCL3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BCL3 - B-cell CLL/lymphoma 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005178.4 | 883 | Intron | NP_005169.2 | |||
XM_011527197.2 | 883 | Intron | XP_011525499.2 | |||
XM_011527198.2 | 883 | Intron | XP_011525500.2 | |||
XM_011527200.2 | 883 | Intron | XP_011525502.1 | |||
XM_017027109.1 | 883 | UTR 5 | XP_016882598.1 | |||
XM_017027110.1 | 883 | UTR 5 | XP_016882599.1 |
Set Membership: |
HapMap |