Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611831 MIM: 186980 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL18 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MRPL18 - mitochondrial ribosomal protein L18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318817.1 | Intron | NP_001305746.1 | ||||
NM_014161.4 | Intron | NP_054880.2 |
PNLDC1 - PARN like, ribonuclease domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA29 - small nucleolar RNA, H/ACA box 29 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TCP1 - t-complex 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008897.1 | Intron | NP_001008897.1 | ||||
NM_030752.2 | Intron | NP_110379.2 |
Set Membership: |
HapMap |