Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAATACCTGGAGAATTACCTCAAC[C/T]GTCTCTTGACCATGTCTTTCTATCG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602384 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107984973 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC107984973 - atherin-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLD2 - phospholipase D2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243108.1 | 589 | Missense Mutation | CGT,TGT | R,C 172 | NP_001230037.1 | |
NM_002663.4 | 589 | Missense Mutation | CGT,TGT | R,C 172 | NP_002654.3 | |
XM_005256696.2 | 589 | Intron | XP_005256753.1 | |||
XM_017024764.1 | 589 | UTR 5 | XP_016880253.1 |
Set Membership: |
HapMap |