Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
52 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616072 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HP1BP3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HP1BP3 - heterochromatin protein 1 binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016287.3 | Intron | NP_057371.2 | ||||
XM_005245875.4 | Intron | XP_005245932.1 | ||||
XM_005245876.3 | Intron | XP_005245933.1 | ||||
XM_005245877.4 | Intron | XP_005245934.1 | ||||
XM_005245878.4 | Intron | XP_005245935.1 | ||||
XM_005245879.4 | Intron | XP_005245936.1 | ||||
XM_011541532.1 | Intron | XP_011539834.1 | ||||
XM_011541533.1 | Intron | XP_011539835.1 | ||||
XM_011541534.2 | Intron | XP_011539836.1 | ||||
XM_017001393.1 | Intron | XP_016856882.1 | ||||
XM_017001394.1 | Intron | XP_016856883.1 |
Set Membership: |
HapMap JSNP |