Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGGCCATAATATATCTTCACACA[C/T]GCTTCCTTCTTTCTGATGCCATTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BZW1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
BZW1 - basic leucine zipper and W2 domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207067.1 | Intron | NP_001193996.1 | ||||
NM_001207069.1 | Intron | NP_001193998.1 | ||||
NM_001321688.1 | Intron | NP_001308617.1 | ||||
NM_001321690.1 | Intron | NP_001308619.1 | ||||
NM_001321691.1 | Intron | NP_001308620.1 | ||||
NM_001321693.1 | Intron | NP_001308622.1 | ||||
NM_001321694.1 | Intron | NP_001308623.1 | ||||
NM_014670.3 | Intron | NP_055485.2 | ||||
XM_017005393.1 | Intron | XP_016860882.1 |
LOC101927795 - uncharacterized LOC101927795 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |