Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606557 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCL11A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
BCL11A - B-cell CLL/lymphoma 11A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018014.3 | 3678 | Intron | NP_060484.2 | |||
NM_022893.3 | 3678 | UTR 3 | NP_075044.2 | |||
NM_138559.1 | 3678 | Intron | NP_612569.1 | |||
XM_011532909.1 | 3678 | Intron | XP_011531211.1 | |||
XM_011532910.1 | 3678 | Intron | XP_011531212.1 | |||
XM_011532912.1 | 3678 | Intron | XP_011531214.1 | |||
XM_017004333.1 | 3678 | UTR 3 | XP_016859822.1 | |||
XM_017004334.1 | 3678 | UTR 3 | XP_016859823.1 | |||
XM_017004335.1 | 3678 | UTR 3 | XP_016859824.1 | |||
XM_017004336.1 | 3678 | UTR 3 | XP_016859825.1 | |||
XM_017004337.1 | 3678 | UTR 3 | XP_016859826.1 | |||
XM_017004338.1 | 3678 | UTR 3 | XP_016859827.1 |
Set Membership: |
HapMap |