Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603564 MIM: 610891 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DPM2 - dolichyl-phosphate mannosyltransferase polypeptide 2, regulatory subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM102A - family with sequence similarity 102 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035254.2 | 892 | Silent Mutation | GAC,GAT | D,D 338 | NP_001030331.1 | |
NM_203305.2 | 892 | Silent Mutation | GAC,GAT | D,D 196 | NP_976050.1 |