Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
47 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610480 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LEMD1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LEMD1 - LEM domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001552.4 | 561 | Intron | NP_001001552.3 | |||
NM_001199050.1 | 561 | Missense Mutation | GGG,TGG | G,W 91 | NP_001185979.1 | |
NM_001199051.1 | 561 | Missense Mutation | GGG,TGG | G,W 50 | NP_001185980.1 | |
NM_001199052.1 | 561 | Intron | NP_001185981.1 | |||
XM_011510160.2 | 561 | Missense Mutation | GGG,TGG | G,W 119 | XP_011508462.1 | |
XM_011510161.2 | 561 | Missense Mutation | GGG,TGG | G,W 119 | XP_011508463.1 | |
XM_011510162.2 | 561 | Missense Mutation | GGG,TGG | G,W 113 | XP_011508464.1 | |
XM_011510163.2 | 561 | Missense Mutation | GGG,TGG | G,W 78 | XP_011508465.1 | |
XM_011510164.2 | 561 | Missense Mutation | CAG,CAT | Q,H 134 | XP_011508466.1 | |
XM_011510165.2 | 561 | Silent Mutation | CGG,CGT | R,R 126 | XP_011508467.1 |
LEMD1-AS1 - LEMD1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap JSNP |