Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
27 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR7856 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR7856 - microRNA 7856 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ODF2L - outer dense fiber of sperm tails 2 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007022.2 | 2965 | Intron | NP_001007023.2 | |||
NM_001184765.1 | 2965 | UTR 3 | NP_001171694.1 | |||
NM_001184766.1 | 2965 | UTR 3 | NP_001171695.1 | |||
NM_020729.2 | 2965 | Intron | NP_065780.2 | |||
XM_005271054.3 | 2965 | UTR 3 | XP_005271111.1 | |||
XM_011541815.2 | 2965 | UTR 3 | XP_011540117.1 | |||
XM_011541819.2 | 2965 | Intron | XP_011540121.1 | |||
XM_011541821.2 | 2965 | Intron | XP_011540123.1 | |||
XM_011541823.2 | 2965 | Intron | XP_011540125.1 | |||
XM_011541824.2 | 2965 | Intron | XP_011540126.1 | |||
XM_017001876.1 | 2965 | UTR 3 | XP_016857365.1 | |||
XM_017001877.1 | 2965 | UTR 3 | XP_016857366.1 | |||
XM_017001878.1 | 2965 | UTR 3 | XP_016857367.1 | |||
XM_017001879.1 | 2965 | Intron | XP_016857368.1 | |||
XM_017001880.1 | 2965 | UTR 3 | XP_016857369.1 | |||
XM_017001881.1 | 2965 | UTR 3 | XP_016857370.1 | |||
XM_017001882.1 | 2965 | UTR 3 | XP_016857371.1 |