Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTCTTTTCAAACTTGCACGTTGG[A/G]GTTTTAAACACAACCACTTTACATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606108 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927702 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101927702 - uncharacterized LOC101927702 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPM1A - protein phosphatase, Mg2+/Mn2+ dependent 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021003.4 | Intron | NP_066283.1 | ||||
NM_177951.2 | Intron | NP_808820.1 | ||||
NM_177952.2 | Intron | NP_808821.2 | ||||
XM_005267779.1 | Intron | XP_005267836.1 | ||||
XM_005267781.1 | Intron | XP_005267838.1 | ||||
XM_011536872.1 | Intron | XP_011535174.1 | ||||
XM_011536876.1 | Intron | XP_011535178.1 | ||||
XM_011536877.1 | Intron | XP_011535179.1 | ||||
XM_011536878.1 | Intron | XP_011535180.1 | ||||
XM_011536879.2 | Intron | XP_011535181.1 | ||||
XM_011536880.2 | Intron | XP_011535182.1 | ||||
XM_011536881.2 | Intron | XP_011535183.1 | ||||
XM_011536882.2 | Intron | XP_011535184.1 | ||||
XM_011536883.1 | Intron | XP_011535185.1 | ||||
XM_011536884.1 | Intron | XP_011535186.1 | ||||
XM_017021381.1 | Intron | XP_016876870.1 | ||||
XM_017021382.1 | Intron | XP_016876871.1 | ||||
XM_017021383.1 | Intron | XP_016876872.1 | ||||
XM_017021384.1 | Intron | XP_016876873.1 | ||||
XM_017021385.1 | Intron | XP_016876874.1 | ||||
XM_017021386.1 | Intron | XP_016876875.1 | ||||
XM_017021387.1 | Intron | XP_016876876.1 |