Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
![]() |
||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF234 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF234 - zinc finger protein 234 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144824.1 | 464 | Missense Mutation | ATC,GTC | I,V 16 | NP_001138296.1 | |
NM_006630.2 | 464 | Missense Mutation | ATC,GTC | I,V 16 | NP_006621.1 | |
XM_006722974.3 | 464 | Missense Mutation | ATC,GTC | I,V 16 | XP_006723037.1 | |
XM_017026149.1 | 464 | Missense Mutation | ATC,GTC | I,V 16 | XP_016881638.1 | |
XM_017026150.1 | 464 | Missense Mutation | ATC,GTC | I,V 16 | XP_016881639.1 | |
XM_017026151.1 | 464 | Missense Mutation | ATC,GTC | I,V 16 | XP_016881640.1 | |
XM_017026152.1 | 464 | UTR 5 | XP_016881641.1 | |||
XM_017026153.1 | 464 | UTR 5 | XP_016881642.1 |
Set Membership: |
![]() |