Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGATTTGTTACAGCAGTTGCATC[A/G]TAGGAAGGGGTGACTTTAGAATAAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616066 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC169-SOHLH2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC169-SOHLH2 - CCDC169-SOHLH2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198910.1 | 1671 | Silent Mutation | TAC,TAT | Y,Y 452 | NP_001185839.1 |
SOHLH2 - spermatogenesis and oogenesis specific basic helix-loop-helix 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282147.1 | 1671 | Intron | NP_001269076.1 | |||
NM_017826.2 | 1671 | Silent Mutation | TAC,TAT | Y,Y 375 | NP_060296.2 |
Set Membership: |
HapMap |