Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608550 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCED1A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PCED1A - PC-esterase domain containing 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271168.1 | 289 | UTR 5 | NP_001258097.1 | |||
NM_022760.4 | 289 | Intron | NP_073597.2 | |||
XM_005260804.2 | 289 | UTR 5 | XP_005260861.1 | |||
XM_005260805.2 | 289 | Intron | XP_005260862.1 | |||
XM_005260806.3 | 289 | Intron | XP_005260863.1 |
VPS16 - VPS16, CORVET/HOPS core subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022575.3 | 289 | Intron | NP_072097.2 | |||
NM_080413.2 | 289 | Intron | NP_536338.1 |
Set Membership: |
HapMap Validated |