Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606421 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELMO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ELMO2 - engulfment and cell motility 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318253.1 | Intron | NP_001305182.1 | ||||
NM_133171.4 | Intron | NP_573403.1 | ||||
NM_182764.2 | Intron | NP_877496.1 | ||||
XM_005260496.3 | Intron | XP_005260553.1 | ||||
XM_005260498.3 | Intron | XP_005260555.1 | ||||
XM_005260499.3 | Intron | XP_005260556.1 | ||||
XM_005260500.3 | Intron | XP_005260557.1 | ||||
XM_005260501.4 | Intron | XP_005260558.1 | ||||
XM_006723854.3 | Intron | XP_006723917.1 | ||||
XM_017028009.1 | Intron | XP_016883498.1 | ||||
XM_017028010.1 | Intron | XP_016883499.1 | ||||
XM_017028011.1 | Intron | XP_016883500.1 |
SLC35C2 - solute carrier family 35 member C2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |