Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCTTCCCTGCCCGTCCCCCCCAC[C/T]GGGTGGAGATCAGGAGCCCAGCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300708 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM58A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM58A - family with sequence similarity 58 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130997.2 | Intron | NP_001124469.1 | ||||
NM_152274.4 | Intron | NP_689487.2 | ||||
XM_005277920.4 | Intron | XP_005277977.1 | ||||
XM_005277921.4 | Intron | XP_005277978.1 | ||||
XM_011531214.2 | Intron | XP_011529516.1 | ||||
XM_011531215.2 | Intron | XP_011529517.1 |
LOC105373383 - uncharacterized LOC105373383 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |