Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613486 MIM: 607642 MIM: 184756 | ||||||||||||||||||||
Literature Links: |
MIR33B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR33B - microRNA 33b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6777 - microRNA 6777 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAI1 - retinoic acid induced 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SREBF1 - sterol regulatory element binding transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005291.2 | 3384 | Missense Mutation | CGG,TGG | R,W 1094 | NP_001005291.1 | |
NM_001321096.2 | 3384 | Missense Mutation | CGG,TGG | R,W 1040 | NP_001308025.1 | |
NM_004176.4 | 3384 | Missense Mutation | CGG,TGG | R,W 1064 | NP_004167.3 | |
XM_005256772.4 | 3384 | Missense Mutation | CGG,TGG | R,W 1062 | XP_005256829.1 | |
XM_017024970.1 | 3384 | Missense Mutation | CGG,TGG | R,W 683 | XP_016880459.1 | |
XM_017024971.1 | 3384 | Missense Mutation | CGG,TGG | R,W 583 | XP_016880460.1 |