Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCAGGCTCTTTTCTGACTTTGTC[A/G]AATCACATTTTTGCAGGTGAATGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
64 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602600 MIM: 602603 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100507564 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100507564 - uncharacterized LOC100507564 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRP8 - LDL receptor related protein 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAGOH - mago homolog, exon junction complex core component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002370.3 | Intron | NP_002361.1 |