Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608121 MIM: 608122 | ||||||||||||||||||||
Literature Links: |
PARVB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PARVB - parvin beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PARVG - parvin gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001137605.2 | 2631 | Intron | NP_001131077.1 | |||
NM_022141.6 | 2631 | Intron | NP_071424.1 | |||
XM_005261702.3 | 2631 | Intron | XP_005261759.1 | |||
XM_011530302.2 | 2631 | UTR 5 | XP_011528604.1 | |||
XM_017028907.1 | 2631 | Intron | XP_016884396.1 | |||
XM_017028908.1 | 2631 | Intron | XP_016884397.1 |