Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
24 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605557 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRDM16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRDM16 - PR domain 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022114.3 | Intron | NP_071397.3 | ||||
NM_199454.2 | Intron | NP_955533.2 | ||||
XM_005244772.4 | Intron | XP_005244829.1 | ||||
XM_005244773.4 | Intron | XP_005244830.1 | ||||
XM_005244774.4 | Intron | XP_005244831.1 | ||||
XM_006710814.3 | Intron | XP_006710877.1 | ||||
XM_011541945.2 | Intron | XP_011540247.1 | ||||
XM_017002050.1 | Intron | XP_016857539.1 |