Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
25 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609269 MIM: 605764 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA0319 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EUR
|
African American - Not Available | YRI (Yoruba)
|
||||||
EAS - Not Available | Japanese
|
JPT (Japanese)
|
||||||
SAS - Not Available | Chinese
|
CHB (Han Chinese)
|
||||||
AFR - Not Available | ||||||||
AMR
|
KIAA0319 - KIAA0319 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TDP2 - tyrosyl-DNA phosphodiesterase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016614.2 | Intron | NP_057698.2 |