Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606749 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TINAG PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
TINAG - tubulointerstitial nephritis antigen | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014464.3 | 211 | Missense Mutation | CAG,CGG | Q,R 22 | NP_055279.3 | |
XM_006715062.2 | 211 | Missense Mutation | CAG,CGG | Q,R 4 | XP_006715125.1 | |
XM_011514497.2 | 211 | Missense Mutation | CAG,CGG | Q,R 22 | XP_011512799.1 | |
XM_017010745.1 | 211 | Missense Mutation | CAG,CGG | Q,R 22 | XP_016866234.1 | |
XM_017010746.1 | 211 | Missense Mutation | CAG,CGG | Q,R 22 | XP_016866235.1 | |
XM_017010747.1 | 211 | Missense Mutation | CAG,CGG | Q,R 22 | XP_016866236.1 | |
XM_017010748.1 | 211 | Intron | XP_016866237.1 |
Set Membership: |
HapMap |