Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
26 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606351 MIM: 611101 MIM: 603366 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESPN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ESPN - espin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLEKHG5 - pleckstrin homology and RhoGEF domain containing G5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042663.1 | 4331 | UTR 3 | NP_001036128.1 | |||
NM_001042664.1 | 4331 | UTR 3 | NP_001036129.1 | |||
NM_001042665.1 | 4331 | UTR 3 | NP_001036130.1 | |||
NM_001265592.1 | 4331 | UTR 3 | NP_001252521.1 | |||
NM_001265593.1 | 4331 | UTR 3 | NP_001252522.1 | |||
NM_001265594.1 | 4331 | UTR 3 | NP_001252523.1 | |||
NM_020631.4 | 4331 | UTR 3 | NP_065682.2 | |||
NM_198681.3 | 4331 | UTR 3 | NP_941374.2 |
TNFRSF25 - TNF receptor superfamily member 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039664.1 | 4331 | Intron | NP_001034753.1 | |||
NM_003790.2 | 4331 | Intron | NP_003781.1 | |||
NM_148965.1 | 4331 | Intron | NP_683866.1 | |||
NM_148966.1 | 4331 | Intron | NP_683867.1 | |||
NM_148967.1 | 4331 | Intron | NP_683868.1 | |||
NM_148970.1 | 4331 | Intron | NP_683871.1 |
Set Membership: |
HapMap |