Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
70 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604128 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCEA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TCEA3 - transcription elongation factor A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003196.2 | 1244 | Intron | NP_003187.1 | |||
XM_006710864.1 | 1244 | UTR 3 | XP_006710927.1 | |||
XM_011542053.2 | 1244 | Intron | XP_011540355.1 | |||
XM_017002200.1 | 1244 | Missense Mutation | AGT,GGT | S,G 385 | XP_016857689.1 | |
XM_017002201.1 | 1244 | Missense Mutation | AGT,GGT | S,G 364 | XP_016857690.1 | |
XM_017002202.1 | 1244 | Missense Mutation | AGT,GGT | S,G 364 | XP_016857691.1 | |
XM_017002203.1 | 1244 | Missense Mutation | AGT,GGT | S,G 348 | XP_016857692.1 | |
XM_017002204.1 | 1244 | Intron | XP_016857693.1 |
Set Membership: |
HapMap JSNP |