Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
38 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608198 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PGLYRP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PGLYRP4 - peptidoglycan recognition protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020393.3 | Intron | NP_065126.2 | ||||
XM_011509789.2 | Intron | XP_011508091.1 | ||||
XM_011509790.1 | Intron | XP_011508092.1 | ||||
XM_011509791.2 | Intron | XP_011508093.1 | ||||
XM_011509792.1 | Intron | XP_011508094.1 | ||||
XM_011509793.2 | Intron | XP_011508095.1 |